Learn More
Biomatik Corporation Mouse Monocyte Chemotactic Protein 2 (MCP2) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11228
Encompass_Preferred
Mouse Monocyte Chemotactic Protein 2 (MCP2) Mini sample ELISA Kit for detecting MCP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z121. Gene ID: 20307. Target Synonyms: CCL8; CCL-8; SCYA8; MCP-2; HC14; Chemokine C-C-Motif Ligand 8; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 8; Monocyte Chemoattractant Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4555