Learn More
Biomatik Corporation Mouse Monokine Induced By Interferon Gamma (MIg) CLIA Kit
Supplier:  Biomatik Corporation EKU09703
Encompass_Preferred
Mouse Monokine Induced By Interferon Gamma (MIg) CLIA Kit for detecting MIg in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.12-3,000pg/ml. Sensitivity: 1.96pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P18340. Gene ID: 17329. Target Synonyms: CXCL9; CMK; Humig; MI-G; SCYB9; Crg-10; Chemokine (C-X-C motif) ligand 9; Small-inducible cytokine B9; Gamma-interferon-induced monokine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5967