Learn More
Biomatik Corporation Mouse Mothers Against Decapentaplegic Homolog 2 (Smad2) ELISA Kit
Supplier:  Biomatik Corporation EKU11294
Encompass_Preferred
Mouse Mothers Against Decapentaplegic Homolog 2 (Smad2) ELISA Kit for detecting Smad2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 28.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62432. Gene ID: 17126. Target Synonyms: JV18; JV18-1; MADH2; MADR2; HMAD-2; HSMAD2; Mad-related protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6157