Learn More
BIOHIPPO Mouse MPO Microsample
SDP
Catalog No. 5000314770
Encompass
Mouse MPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MPO in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 10 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14770 Supplier BIOHIPPO Supplier No. BHE15212646
Mouse MPO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MPO in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 10 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MPO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MPO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MPO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color