Learn More
Biomatik Corporation Mouse αMSH (Alpha-Melanocyte Stimulating Hormone) ELISA Kit
Supplier:  Biomatik Corporation EKF58485
Encompass_Preferred
Mouse αMSH (Alpha-Melanocyte Stimulating Hormone) ELISA Kit for detecting αMSH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300pg/ml. Sensitivity: 2.813pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01193. Target Synonyms: αMSH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9176