Learn More
BIOHIPPO Mouse MSP Microsample
SDP
Catalog No. 5000314737
Encompass
Mouse MSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSP in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14737 Supplier BIOHIPPO Supplier No. BHE15212537
Mouse MSP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MSP in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 056 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MSP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MSP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MSP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color