Learn More
Sino Biological Mouse MSR1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50129R004100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MSR1 / CD204 (rM MSR1 / CD204; Catalog#50129-M07H; AAH03814.1; Trp 83-Val 354).
Catalog No. 50-237-4004