Learn More
Sino Biological Recombinant Anti-CD204/MSR1 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 50129R004F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse MSR1 / CD204 (rM MSR1 / CD204; Catalog#50129-M07H; AAH03814.1; Trp 83-Val 354) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8926