Learn More
Sino Biological Anti-CD204/MSR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50129RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse MSR1 / CD204 (rM MSR1 / CD204; Catalog#50129-M07H; AAH03814.1; Trp 83-Val 354). MSR1 / CD204 specific IgG was purified by Mouse MSR1 / CD204 affinity chromatography.
Catalog No. 50-240-8150