Learn More
BIOHIPPO Mouse mTOR Microsample
SDP
Catalog No. 5000314807
Encompass
Mouse mTOR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of mTOR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 131 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MTOR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MTOR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MTOR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14807 Supplier BIOHIPPO Supplier No. BHE15210664
Mouse mTOR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of mTOR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 131 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MTOR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MTOR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MTOR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color