Learn More
BIOHIPPO Mouse MUC5B Microsample
SDP
Catalog No. 5000314176
Encompass
Mouse MUC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC5B in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 247 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MUC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MUC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MUC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14176 Supplier BIOHIPPO Supplier No. BHE15212273
Mouse MUC5B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC5B in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 247 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MUC5B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MUC5B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MUC5B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color