Learn More
Biomatik Corporation Mouse Multimerin 2 (MMRN2) ELISA Kit
Supplier:  Biomatik Corporation EKU06042
Encompass_Preferred
Mouse Multimerin 2 (MMRN2) ELISA Kit for detecting MMRN2 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A6H6E2. Gene ID: 105450. Target Synonyms: EMILIN3; EndoGlyx-1; Elastin Microfibril Interfacer 3; Elastin microfibril interface located protein 3; EndoGlyx-1 p125/p140 subunit. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8860