Learn More
Biomatik Corporation Mouse Myeloid Differentiation Factor 88 (MyD88) ELISA Kit
Supplier:  Biomatik Corporation EKU06060
Encompass_Preferred
Mouse Myeloid Differentiation Factor 88 (MyD88) ELISA Kit for detecting MyD88 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22366. Gene ID: 17874. Target Synonyms: Myeloid Differentiation Primary Response Gene 88. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7032