Learn More
Biomatik Corporation Mouse Myeloid Progenitor Inhibitory Factor 2 (MPIF2) CLIA Kit
Supplier:  Biomatik Corporation EKU08910
Encompass_Preferred
Mouse Myeloid Progenitor Inhibitory Factor 2 (MPIF2) CLIA Kit for detecting MPIF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.97pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9JKC0. Gene ID: 56221. Target Synonyms: CCL24; SCYA24; Ckb-6; Eotaxin-2; Chemokine C-C-Motif Ligand 24; Eosinophil chemotactic protein 2; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6773