Learn More
American Research Products Inc Mouse myeloperoXidase antineut
Catalog No. 5000329495
Encompass
Mouse myeloperoXidase-antineutrophil cytoplasmic antibody ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29495 Supplier American Research Products Inc Supplier No. CSBE08676M
Mouse myeloperoXidase-antineutrophil cytoplasmic antibody ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma Method Sandwich