Learn More
Biomatik Corporation Mouse Myeloperoxidase (MPO) CLIA Kit
Supplier:  Biomatik Corporation EKU08903
Encompass_Preferred
Mouse Myeloperoxidase (MPO) CLIA Kit for detecting MPO in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P11247. Gene ID: 17523. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5397