Learn More
BIOHIPPO Mouse MYH2 Microsample
SDP
Catalog No. 5000315546
Encompass
Mouse MYH2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH2 in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 049 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MYH2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MYH2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MYH2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15546 Supplier BIOHIPPO Supplier No. BHE15212631
Mouse MYH2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH2 in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 049 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MYH2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MYH2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MYH2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color