Learn More
BIOHIPPO Mouse MYO Microsample
SDP
Catalog No. 5000314212
Encompass
Mouse MYO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYO in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 0 86 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MYO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MYO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MYO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14212 Supplier BIOHIPPO Supplier No. BHE15212598
Mouse MYO Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYO in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 0 86 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse MYO Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse MYO Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse MYO biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color