Learn More
Biomatik Corporation Mouse Myogenic Differentiation (MyoD) ELISA Kit
Supplier:  Biomatik Corporation EKU12324
Encompass_Preferred
Mouse Myogenic Differentiation (MyoD) ELISA Kit for detecting MyoD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10085. Gene ID: 17927. Target Synonyms: MYOD1; PUM; MYF3; bHLHc1; Myoblast Determination Protein 1; Myogenic Factor 3; Class C basic helix-loop-helix protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7333