Learn More
Biomatik Corporation Mouse Myoglobin (MYO) CLIA Kit
Supplier:  Biomatik Corporation EKU08871
Encompass_Preferred
Mouse Myoglobin (MYO) CLIA Kit for detecting MYO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 2.14pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04247. Gene ID: 17189. Target Synonyms: PVALB; MB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6038