Learn More
Biomatik Corporation Mouse Myosin Heavy Chain 1 (MYH1) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU11533
Encompass_Preferred
Mouse Myosin Heavy Chain 1 (MYH1) Wide-range ELISA Kit for detecting MYH1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.131ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5SX40. Gene ID: 17879. Target Synonyms: MYHSA1; MYHa; MyHC-2X/D; Myosin-1; Myosin Heavy Chain IIx/d; Myosin Heavy Chain 1, Skeletal Muscle, Adult. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7514