Learn More
Biomatik Corporation Mouse Myosin Heavy Chain 4, Skeletal Muscle (MYH4) ELISA Kit
Supplier:  Biomatik Corporation EKU06095
Encompass_Preferred
Mouse Myosin Heavy Chain 4, Skeletal Muscle (MYH4) ELISA Kit for detecting MYH4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5SX39. Gene ID: 17884. Target Synonyms: MYH2B; Myosin heavy chain 2b; Myosin heavy chain, skeletal muscle, fetal. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8857