Learn More
American Research Products Inc Mouse N myc interactor NMI E
Catalog No. 5000328120
Encompass
Mouse N-myc-interactor (NMI) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28120 Supplier American Research Products Inc Supplier No. CSBEL015893MO
Mouse N-myc-interactor (NMI) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich