Learn More
Sino Biological Recombinant Anti-NAALADL1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50937R005100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse NAALADL1 (Catalog#50937-M07H; Q7M758; Pro29-Leu745).
Catalog No. 50-241-1814