Learn More
Biomatik Corporation Mouse N-Acetyltransferase 1 (NAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU06136
Encompass_Preferred
Mouse N-Acetyltransferase 1 (NAT1) ELISA Kit for detecting NAT1 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50294. Gene ID: 17960. Target Synonyms: AAC1; Arylamine N-Acetyltransferase; Arylamide acetylase 1; Monomorphic arylamine N-acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4618