Learn More
American Research Products Inc Mouse NAD P H dehydrogenase
Catalog No. 5000327515
Encompass
Mouse NAD (P) H dehydrogenase [quinone] 1 (NQO1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27515 Supplier American Research Products Inc Supplier No. CSBEL016039MO
Mouse NAD (P) H dehydrogenase [quinone] 1 (NQO1) ELISA kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich