Learn More
Biomatik Corporation Mouse NADH Dehydrogenase 1 (ND1) ELISA Kit
Supplier:  Biomatik Corporation EKU06143
Encompass_Preferred
Mouse NADH Dehydrogenase 1 (ND1) ELISA Kit for detecting ND1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P03888. Gene ID: 17716. Target Synonyms: MT-ND1; MTND1; NAD1; NADH-ubiquinone oxidoreductase chain 1; Mitochondrially Encoded NADH Dehydrogenase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6629