Learn More
ACROBiosystems Mouse Nectin-4 25ug
Catalog No. 502534383
Encompass_Preferred
Biotinylated Mouse Nectin-4 Protein HisAvitag (NE4-M82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1).
Catalog No. 50-253-4383 Supplier ACROBiosystems Supplier No. NE4M82E725UG
Biotinylated Mouse Nectin-4 Protein HisAvitag (NE4-M82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1).