Learn More
ACROBiosystems Mouse Nectin-4 Protein, Fc Tag
Supplier:  ACROBiosystems NE4M52571MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Nectin-4, Fc Tag (NE4-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.8 kDa.
Catalog No. 50-205-4882