Learn More
ACROBiosystems Mouse Nectin-4 Protein, Fc Tag
Supplier: ACROBiosystems NE4M5257100UG
Recombinant Protein;100UG;Mouse Nectin-4, Fc Tag (NE4-M5257) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession # Q8R007-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.8 kDa.