Learn More
Sino Biological Recombinant Mouse Nectin-1/PVRL1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 58137M08H100
Encompass_Preferred
A DNA sequence encoding the mouse NECTIN1 (NP_067399.2) (Met1-Ala354) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2944