Learn More
Sino Biological Recombinant Mouse Nectin-4 Protein (ECD, His Tag)
Supplier:  Sino Biological 5A7674M08H100
Encompass_Preferred
A DNA sequence encoding the Mouse NECTIN4 (Q8R007-1) (Met1-Ser347) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6543