Learn More
Biomatik Corporation Mouse Nei Endonuclease VIII Like Protein 3 (NEIL3) ELISA Kit
Supplier:  Biomatik Corporation EKU06160
Encompass_Preferred
Mouse Nei Endonuclease VIII Like Protein 3 (NEIL3) ELISA Kit for detecting NEIL3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K203. Gene ID: 234258. Target Synonyms: FPG2; hFPG2; hNEI3; Endonuclease 8-like 3; DNA glycosylase FPG2; DNA glycosylase/AP lyase Neil3; Nei-like protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7029