Learn More
World Precision Instrument WPI Standard Mouse & Neonatal Rat Dual Stereotaxic Instrument With Adaptors
SDP
Catalog No. 502539179
Encompass
Using model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The Mouse/Neonatal Rat Adaptor is designed for use with the traditional 'U' frame Stereotaxic instrument. The adaptor employs light, Delrin Ear Bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9179 Supplier World Precision Instrument Supplier No. 505236
Add to Cart
Add to Cart
Request a Quote
Using model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The Mouse/Neonatal Rat Adaptor is designed for use with the traditional 'U' frame Stereotaxic instrument. The adaptor employs light, Delrin Ear Bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups.
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart