Learn More
World Precision Instrument Mouse & Neonatal Rat Motor
SDP
Catalog No. 502539077
Encompass
Motorized Stereotaxic frames are designed for fast, accurate positioning during stereotaxic surgery. Ideal for inexperienced users, the motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis. Manual movement available anytime. Optional Drill feature available 505379.
Catalog No. 50-253-9077 Supplier World Precision Instrument Supplier No. 505381
Add to Cart
Add to Cart
Request a Quote
Motorized Stereotaxic frames are designed for fast, accurate positioning during stereotaxic surgery. Ideal for inexperienced users, the motorized manipulator arms employ a touchscreen display for easy-to-control movement of each axis. Manual movement available anytime. Optional Drill feature available 505379.