Learn More
Biomatik Corporation Mouse Nephrin (NPHN) CLIA Kit
Supplier:  Biomatik Corporation EKU08928
Encompass_Preferred
Mouse Nephrin (NPHN) CLIA Kit for detecting NPHN in urine, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9QZS7. Gene ID: 54631. Target Synonyms: NPHS1; CNF; Nephrosis 1; Congenital Finnish Type; Renal glomerulus-specific cell adhesion receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7251