Learn More
American Research Products Inc Mouse NerVe growth factor NGF
Catalog No. 5000329882
Encompass
Mouse NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29882 Supplier American Research Products Inc Supplier No. CSBE04684M
Mouse NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 62 5 pg/ml-4000 pg/ml Sensitivity 15 6 pg/ml Sample serum plasma tissue homogenates Method Sandwich