Learn More
Biomatik Corporation Mouse Nerve Growth Factor (NGF) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11234
Encompass_Preferred
Mouse Nerve Growth Factor (NGF) Mini sample ELISA Kit for detecting NGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6LDU8. Gene ID: 18049. Target Synonyms: NGFB; Beta-NGF; HSAN5; NGF-B; Beta-Nerve Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8191