Learn More
Biomatik Corporation Mouse Nesfatin 1 (NES1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11615
Encompass_Preferred
Mouse Nesfatin 1 (NES1) Mini sample ELISA Kit for detecting NES1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 493.8-40,000pg/ml. Sensitivity: 187.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81117. Gene ID: 53322. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6898