Learn More
R&D Systems™ Mouse Netrin-3 (NP_035077) VersaClone cDNA
Description
- Also known as: Ntn2l
- Nectrin-3 is a member of the netrin family of proteins, which function in various biological processes including axon guidance, tumorigenesis, and angiogenesis.
- Netrins are laminin-related proteins that have an N-terminal laminintype domain, epidermal growth factor-like repeat domain, and a positively charged heparin-binding domain at the C-terminus.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mNtn3 |
| Accession Number | NP_035077 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1756 bp |
| Gene ID (Entrez) | 4917 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.