Learn More
Raybiotech Inc C-Series Mouse Neuro Discovery Array C1 Kit.
Supplier:  Raybiotech Inc AAMNEU18
Encompass_Preferred
Detects 23 human neurologically relevant proteins. Suitable for serum, plasma, cell culture media and other liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: Fas Ligand (TNFSF6), Fractalkine (CX3CL1), GCSF, IFN-gamma, IGF-1, IL-10, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-4, IL-6, KC (CXCL1), LIX, MCP-1 (CCL2), M-CSF, MIP-1 alpha (CCL3), MMP-2, MMP-3, RAGE, TARC (CCL17), SDF-1 alpha (CXCL12 alpha), TGF beta 1, TNF alpha, VEGF-A
Catalog No. 50-194-8745