Learn More
Abcam Mouse Neurofilament Heavy Polypeptide ELISA Kit (NEFH), 1 x 96 Tests
Catalog No. 502656017
Encompass_Preferred
Mouse Neurofilament Heavy Polypeptide ELISA Kit (NEFH) is a single-wash 90-min Simplestep used to quantify Mouse Neurofilament Heavy Polypeptide (NEFH) with a sensitivity of 57.12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6017 Supplier Abcam Supplier No. AB2724791X96TESTS
Mouse Neurofilament Heavy Polypeptide ELISA Kit (NEFH) is a single-wash 90-min Simplestep used to quantify Mouse Neurofilament Heavy Polypeptide (NEFH) with a sensitivity of 57.12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement