Learn More
Biomatik Corporation Mouse Neurokinin A (NKA) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12423
Encompass_Preferred
Mouse Neurokinin A (NKA) Mini sample ELISA Kit for detecting NKA in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41539. Gene ID: 21333. Target Synonyms: NK-A; SK; Substance K; Neuropeptide K; Neurokinin K; Neuromedin L; Neuropeptide gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7453