Learn More
Biomatik Corporation Mouse Neuropeptide Y (NPY) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10950
Encompass_Preferred
Mouse Neuropeptide Y (NPY) High-sensitive ELISA Kit for detecting NPY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.47-200pg/ml. Sensitivity: 0.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P57774. Gene ID: 109648. Target Synonyms: NP-Y; PYY4; CPON; Pro-neuropeptide Y; Neuropeptide tyrosine; C-flanking peptide of NPY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8264