Learn More
American Research Products Inc Mouse neutrophil elastase NE
Catalog No. 5000326324
Encompass
Mouse neutrophil elastase NE ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26324 Supplier American Research Products Inc Supplier No. CSBE04804M
Mouse neutrophil elastase NE ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich