Learn More
American Research Products Inc Mouse neutrophil gelatinase as
Catalog No. 5000329911
Encompass
Mouse neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates urine Method Competitive
Catalog No. 50-003-29911 Supplier American Research Products Inc Supplier No. CSBE09410M
Mouse neutrophil gelatinase-associated lipocalin NGAL ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma tissue homogenates urine Method Competitive