Learn More
Immunology Consultants Mouse NGAL ELISA Kit
SDP
Supplier:  Immunology Consultants E90NGL
Encompass_Preferred
The Mouse NGAL test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring NGAL in mouse biological samples.
Catalog No. 50-240-6638