Learn More
FUJIFILM Biosciences Mouse beta-NGF, Recombinant Protein, Shenandoah Biotechnology, 100 ug vial
Catalog No. 50990932
Encompass_Preferred
Nerve growth factor beta (B-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons. B-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways. B-NGF is also involved in the growth, differentiation, and survival of B lymphocytes. Human, mouse, and rat B-NGF proteins are cross-reactive.
Catalog No. 50-990-932 Supplier FUJIFILM Biosciences Supplier No. 20011100UG
Add to Cart
Edge
Add to Cart
Nerve growth factor beta (B-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons. B-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways. B-NGF is also involved in the growth, differentiation, and survival of B lymphocytes. Human, mouse, and rat B-NGF proteins are cross-reactive.