Learn More
Sino Biological Anti-Beta-NGF Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50385RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse NGF / NGFB (Catalog#50385-MNAC; NP_001106168.1; Ser122-Gly241). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9045