Learn More
Sino Biological Mouse NGF PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50385T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse NGF / NGFB (Catalog#50385-MNAC; NP_001106168.1; Ser122-Gly241). NGF / NGFB specific IgG was purified by Mouse NGF / NGFB affinity chromatography.
Catalog No. 50-237-0527